Anti-UTS2 Rabbit Polyclonal Antibody
PRSI19-596
New Product
- Antibody type:Primary
- Antigen name:Urotensin 2
- Antigen symbol:UTS2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:10911
- Antigen synonyms:PRO1068|UII|UCN2|urotensin II|prepro U-II|urotensin-2|U-II
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3.
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 35-124 of human UTS2 (NP_006777.1).
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: UTS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat