Anti-SLC20A1 Rabbit Polyclonal Antibody
Catalog # PRSI19-284
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Solute carrier family 20 (phosphate transporter), member 1
- Antigen symbol:SLC20A1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:6574
- Antigen synonyms:FLJ41426|SLC20A1|GLVR1|Glvr-1|PiT-1|PIT1|DKFZp686J2397
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3.
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 257-356 of human SLC20A1 (NP_005406.3).
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: SLC20A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat