Anti-DRD5 Rabbit Polyclonal Antibody
PRSI18-976
New Product
- Antibody type:Primary
- Antigen name:Dopamine receptor D5
- Antigen symbol:DRD5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Form:Liquid
- Gene ID:1816
- Antigen synonyms:D1beta dopamine receptor|DBDR|DRD1B|DRD1L2|MGC10601|dopamine receptor D1B|dopamine receptor D5|DRD5
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3.
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 361-477 of human DRD5 (NP_000789.1).
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: DRD5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat