Anti-H4R3me1 (methylated) Rabbit Polyclonal Antibody
Catalog # PRSI18-970
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Histone cluster 1, H4a
- Antigen symbol:H4R3me1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:8366
- Antigen synonyms:H4FJ|HIST1H4H|H4/I|H4/M|H4FI|H4/K|H4FA|HIST1H4B|Histone H4H4FA|H4/G|HIST2H4B|HIST1H4E|H4F2|H4/O|HIST1H4L|H4/D|HIST4H4|H4FC|H4/E|H4/B|H4FE|HIST1H4J|H4FH|H4FM|H4FO|H4FB|HIST1H4K|H4/N|H4/J|HIST2H4|H4/C|HIST1H4F|HIST1H4D|H4FG|HIST1H4I|H4/A|H4FK|H4/H|HIST1H4C|H4FN|H4FD|HIST2H4A
- Modification:Methylated
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3.
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:A synthetic peptide of human MonoMethyl-Histone H4-R3.
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: H4R3me1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat