Order Entry
Spain
Orders LinkContactUsLinkComponent
 

Anti-TRIM63 Rabbit Polyclonal Antibody

Vial
  PRSI18-941
Anti-TRIM63 Rabbit Polyclonal Antibody
  PRSI18-941
Anti-TRIM63 Rabbit Polyclonal Antibody

Vial

 
484,00 €
   
1 * 50 µl
 

Type: Primary
Antigen: TRIM63
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

 
  • Pack type:
    Vial
  • Western blot:
    Yes
  • Clonality:
    Polyclonal
  • Purification:
    Affinity purification
  • Antibody type:
    Primary
  • Form:
    Liquid
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Storage temperature:
    Store at −20...+730 °C. Avoid freeze/thaw cycles.
  • Host:
    Rabbit
  • Concentration:
    Batch dependent
  • Shipping temperature:
    20
  • Antigen name:
    Tripartite motif containing 63, E3 ubiquitin protein ligase
  • Cat. No.:
    PRSI18-941
  • Reactivity:
    Human|Rat|Mouse
  • Gene ID:
    84676
  • Antigen synonyms:
    muscle specific ring finger protein 2|MURF1|IRF|FLJ32380|SMRZ|RNF28|iris ring finger protein|striated muscle RING zinc finger protein|MURF2|MURF-1|ring finger protein 28
  • Immunogen:
    A synthetic peptide of human TRIM63.
  • ImmunoChemistry:
    Yes
  • Conjugation:
    Unconjugated
  • Antigen symbol:
    TRIM63
  • Storage buffer:
    PBS with 0,02% sodium azide, 50% glycerol, pH 7,3.