Order Entry
Spain
ContactUsLinkComponent
Anti-MM-302 Recombinant Antibody
Anti-MM-302 Recombinant Antibody
Catalog # PRSI10-924
Supplier:  ProSci Inc.
Anti-MM-302 Recombinant Antibody
Catalog # PRSI10-924
Supplier:  ProSci Inc.

Specifications

  • Antibody type:
    Primary
  • Clonality:
    Recombinant
  • Conjugation:
    Unconjugated
  • Isotype:
    IgG4 kappa
  • Antigen synonyms:
    UCB7858|UCB-7858
  • Modification:
    None
  • Storage buffer:
    PBS buffer pH 7,5
  • Storage temperature:
    Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2…8 °C for 1 - 2 weeks. Store at –20 °C for 12 months. Store at –80 °C for long term storage.
  • Concentration:
    Batch dependent
  • Shipping temperature:
    20
  • Immunogen:
    Chimeric,Humanised/TGM2 [Homo sapiens]
  • Purification:
    >95%
  • Pack type:
    Vial
  • Pk:
    0,1 mg

Specifications

About this item

Type: Primary
Antigen:
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG4-kappa
Reactivity: