Anti-DMRT1 Rabbit Polyclonal Antibody
PRSI60-338
New Product
- Antibody type:Primary
- Antigen name:Doublesex- and mab-3-related transcription factor 1
- Antigen symbol:DMRT1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:296 - 322 aa
- Form:Liquid
- Gene ID:1761
- Modification:None
- Molecular weight:39 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This DMRT1 antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 296 - 322 amino acids from the C-terminal region of human DMRT1.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Pk:400 µl
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: DMRT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 296-322
Host: Rabbit
Isotype: Ig
Reactivity: Human