Anti-SRP14 Rabbit Polyclonal Antibody
Catalog # PRSI60-309
Supplier: ProSci Inc.
Jump to:
Product Details & Documents
For WB starting dilution is: 1:2000.
Type: Primary
Antigen: SRP14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 28-55
Host: Rabbit
Isotype: Ig
Reactivity: Human
Specifications
- Western blot:Yes
- Clonality:Polyclonal
- Modification:None
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Antibody type:Primary
- Epitope:28 - 55 aa
- Form:Liquid
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Host:Rabbit
- Concentration:0,5 mg/ml
- Molecular weight:15 kDa
- Shipping temperature:4 °C
- Antigen name:Signal recognition particle 14 kDa protein
- Cat. No.:PRSI60-309
- Reactivity:Human
- Gene ID:6727
- Immunogen:This SRP14 antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 28 - 55 amino acids from the Central region of human SRP14.
- Conjugation:Unconjugated
- Antigen symbol:SRP14



