Order Entry
Spain
ContactUsLinkComponent
Anti-PRDM9 Recombinant Antibody [Clone: RAB-C370]
Anti-PRDM9 Recombinant Antibody [Clone: RAB-C370]
  PRSI24-849
Anti-PRDM9 Recombinant Antibody [Clone: RAB-C370]
  PRSI24-849

 

  • Antibody type:
    Primary
  • Antigen name:
    PR/SET domain 9
  • Antigen symbol:
    PRDM9
  • Clonality:
    Recombinant
  • Clone:
    RAB-C370
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    Fab fragment kappa
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Gene ID:
    56979
  • Antigen synonyms:
    PR domain zinc finger protein 9|PR domain-containing protein 9|PRDM9-A003-1459|[histone H4]-N-methyl-L-lysine20 N-methyltransferase PRDM9|EC:2.1.1.-|[histone H3]-lysine36 N-trimethyltransferase PRDM9|[histone H4]-lysine20 N-methyltransferase PRDM9|[histone H3]-lysine4 N-trimethyltransferase PRDM9|Histone-lysine N-methyltransferase PRDM9|Protein-lysine N-methyltransferase PRDM9|[histone H3]-lysine9 N-trimethyltransferase PRDM9|anti-PRDM9-RAB-C370
  • Modification:
    Unmodified
  • Storage buffer:
    PBS with 0,02% Proclin 300
  • Storage temperature:
    Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
  • Shipping temperature:
    Shipped on blue ice
  • Immunogen:
    This antibody was obtained by recombinant antibody (rAb) phage display recognizing PRDM9 protein under non-denaturing conditions
  • Pack type:
    Vial
  • Pk:
    0,1 mg

 

 

Type: Primary
Antigen: PR/SET domain 9
Clonality: Recombinant
Clone: RAB-C370
Conjugation: Unconjugated
Host: Recombinant
Isotype: Fab fragment kappa
Reactivity: Human