Anti-Ly6G Recombinant Antibody [Clone: 1A8]
Catalog # PRSI24-299
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:lymphocyte antigen 6 complex, locus G
- Antigen symbol:Ly6g
- Clonality:Recombinant
- Clone:1A8
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Isotype:IgG2a kappa
- Cross adsorption:No
- Form:Liquid
- Gene ID:546644
- Antigen synonyms:y-6G|LGR-1|Lymphocyte antigen 6G|GR1
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:LEW/N rats immunized with 15x10E8 Ly-6G-transfected EL4J cell
- Tested applications:Depletion
- Pack type:Vial
- Pk:0,2 mg
Specifications
About this item
Type: Primary
Antigen: lymphocyte antigen 6 complex, locus G
Clonality: Recombinant
Clone: 1A8
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgG2a kappa
Reactivity: Mouse