Sodium alginate
Supplier: PanReac AppliChem
Certificates
About this item
CAS Number:9005-38-3
:
MDL Number:MFCD00081310
Storage Temperature:Ambient
Frequently Bought Together
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Nitrile gloves, Kimtech™ Opal™
Supplier: KIMBERLY CLARK
VWR®, Nitrile Gloves
Supplier: VWR Collection
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Chemical resistant gloves, AlphaTec® Solvex 37-675
Supplier: Ansell
VWR®, Transfer Pipettes, Graduated
Supplier: VWR Collection
Tris-Acetate-EDTA (TAE) buffer, 50X
Supplier: PanReac AppliChem
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Membrane filters, Millipore Express® PLUS
Supplier: Merck
Quartz microfibre filters for air sampling, grade QM-A, Whatman™
Supplier: Whatman products (Cytiva)
L-Arginine hydrochloride
Supplier: PanReac AppliChem
Zinc oxide ≥99% (metals basis), nanopowder
Supplier: Thermo Fisher Scientific
Sodium alginate
Supplier: Thermo Fisher Scientific
Sodium hydroxide, BAKER, J.T.Baker®
Supplier: Avantor
Tin(II) oxide ≥99%
Supplier: Thermo Fisher Scientific
VWR® Eyepieces
Supplier: VWR Collection
L(+)-Calcium lactate pentahydrate 98%
Supplier: Thermo Fisher Scientific
Alizarin red S (sodium salt)
Supplier: Thermo Fisher Scientific