Order Entry
Spain
ContactUsLinkComponent
Erythromycin (from Streptomyces erythreus) ≥85% (dry basis)
Erythromycin (from Streptomyces erythreus) ≥85% (dry basis)
Catalog # ENZOALX380274G005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Erythromycin (from Streptomyces erythreus) ≥85% (dry basis)
Catalog # ENZOALX380274G005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :733.95 g/mol
Storage Temperature:Freezer
CAS Number:114-07-8
Formula:C₃₇H₆₇NO₁₃
MDL Number:MFCD00084654