Order Entry
Spain
Orders LinkContactUsLinkComponent
Analog Thermostatised Baths
Analog Thermostatised Baths
Certificates
Analog Thermostatised Baths
4 Options Available Below
Product Details & Documents
Thermostatised baths with inner tank made of stainless steel AISI 304 and metallic housing protected with epoxy paint of great resistance to aggressive surroundings.
  • Temperature controlled by means of an hydraulic thermostat, for water and oil
  • Water drainage faucet and handles for a easy transport
Heating resistance embedded made of special alloy of stainless steel, heat and corrosion resistant. Temperature thermostat
synchronised with the 'on' indicator lamp. Safety cut out thermostat with indicator lamp. The unit will cut out if there is insufficient liquid or if the temperature exceeds the maximum admissible temperature.
 : IP 52 protection class.
 : Includes stainless steel cover-resistance.
Product Family Options
Product Information
  • Temperature Range
    Capacity
    Bath W×D×H
    W×D×H
    Weight
    Availability
    Price
  • OVAN10000-01101
    Thermostatised bath, B105E
    Thermostatised bath, B105E
    5...120 °C
    5 L
    300×150×130 mm
    330×180×300 mm
    6,2 kg
    Log in for availability
    Log in for availability
    1 * 1 UN
    1 * 1 UN - 794,00 €
    794,00 €
    1 * 1 UN
       794,00 €
  • OVAN10000-01105
    Thermostatised bath, B205E
    Thermostatised bath, B205E
    5...210 °C
    5 L
    300×150×130 mm
    330×180×300 mm
    6,2 kg
    Log in for availability
    Log in for availability
    1 * 1 UN
    1 * 1 UN - 983,00 €
    983,00 €
    1 * 1 UN
       983,00 €
  • OVAN10000-01106
    Thermostatised bath, B212E
    Thermostatised bath, B212E
    5...210 °C
    12 L
    330×300×130 mm
    360×330×300 mm
    8,5 kg
    Log in for availability
    Log in for availability
    1 * 1 UN
    1 * 1 UN - 1 240,00 €
    1 240,00 €
    1 * 1 UN
       1 240,00 €
  • OVAN10000-01107
    Thermostatised bath, B220E
    Thermostatised bath, B220E
    5...210 °C
    20 L
    500×300×130 mm
    530×330×300 mm
    11,0 kg
    Log in for availability
    Log in for availability
    1 * 1 UN
    1 * 1 UN - 1 280,00 €
    1 280,00 €
    1 * 1 UN
       1 280,00 €
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Tube racks, 24 tubes Ø 20 mm
    Tube racks, 24 tubes Ø 20 mm
    OVAN20000-00166
    Log in for availability
    1 * 1 UN - 133,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Tube racks, 24 tubes Ø 16 mm
    Tube racks, 24 tubes Ø 16 mm
    OVAN20000-00165
    Log in for availability
    1 * 1 UN - 133,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Support for tube racks, for baths of 5 L
    Support for tube racks, for baths of 5 L
    OVAN20000-00161
    Log in for availability
    1 * 1 UN - 55,90 €
Product Information
  • Catalog #
    Availability
    Price
  • Cover, stainless steel, for baths of 5 L
    Cover, stainless steel, for baths of 5 L
    OVAN20000-00105
    Log in for availability
    1 * 1 UN - 224,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Support for tube racks, for baths of 20 L or 2 for baths of 45 L
    Support for tube racks, for baths of 20 L or 2 for baths of 45 L
    OVAN20000-00163
    Log in for availability
    1 * 1 UN - 72,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Cover, stainless steel, for baths of 45 L
    Cover, stainless steel, for baths of 45 L
    OVAN20000-00108
    Log in for availability
    1 * 1 UN - 451,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Cover with 6 disc adapters for baths of 20 L or 2 units for baths of 45 L
    Cover with 6 disc adapters for baths of 20 L or 2 units for baths of 45 L
    OVAN20000-00102
    Log in for availability
    1 * 1 UN - 468,00 €