Anti-PIP1;1, PIP1;2, PIP1;3, PIP1;4, PIP1;5 Rabbit Polyclonal Antibody
AGRIAS224816
: Agrisera
- Antibody type:Primary
- Antigen name:Aquaporins
- Antigen symbol:PIP1;1, PIP1;2, PIP1;3, PIP1;4, PIP1;5
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Arabidopsis
- Format:Lyophilised
- Purity:Antigen affinity purified serum, in PBS pH 7,4
- Storage temperature:Store lyophilised/reconstituted at −20 °C, once reconstituted make aliquots to avoid repeated freeze-thaw cycles.
- Immunogen:KLH-conjugated synthetic peptide derived from N terminus of Raphanus sativus PAQ1O80368. Chosen peptide is conserved in PIP1;1, PIP1;2, PIP1;3 N-terminus of Raphanus sativus and in all 5 isoforms of Arabidopsis thaliana coded by:AT3G61430.1 (PIP1;1), AT2G45960.3 (PIP1;2), AT1G01620.1 (PIP1;3), AT4G00430.1 (PIP1;4), AT4G23400.1 (PIP1;5)
- Tested applications:Western blot (WB)
- Pk:50 µG
PIPs proteins are aquaporins which facilitate the transport of water and small neutral molecules across cell membrane. PIP1;2 is also designated as PIP1B.
- For reconstitution add 50 µl, of sterile or deionised water
Spin tubes briefly prior to opening them to avoid any losses that might occur from lyophilised material adhering to the cap or sides of the tubes.
Recommended dilution: 1:1000 (WB).
Expected molecular weight: 30,66 kDa
Apparent molecular weight: 28 kDa
: For research use only.
Type: Primary
Antigen: PIP1;1, PIP1;2, PIP1;3, PIP1;4, PIP1;5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Arabidopsis thaliana