o-Tolidina
Catalog #
Product Details & Documents
Synonyms: 4,4'-Bi-o-toluidina
- CAS Number:119-93-7
- Formula:[-C₆H₃(CH₃)-₄-NH₂]₂
- MDL Number:MFCD00014773
- Molecular Weight:212.29 g/mol
- Merck Index:12,09651
- Boiling Pt:∼361 °C (1013 hPa)
- Melting Pt:129…131 °C
- Density:∼1,234 g/cm³ (20 °C)
- Storage Temperature:Ambiente
- EINECS:204-358-0
- UN:3077
- ADR:9,III



