DURAN® Sealing Rings, for Polished Sperical Joints (VITON)
Ball joints without ground joints only seal via the sealing ring.
- Non autoclavable
- Used in vacuum applications
Due to the material properties, it can be used for work with high demands on the chemical resistance of the sealing ring.
Frequently Bought Together