Order Entry
Spain
ContactUsLinkComponent
Certified Reference Material for Standardisation of Volumetric Solutions
Certified Reference Material for Standardisation of Volumetric Solutions
Catalog # ACSDVS66332.80G
Supplier:  CPACHEM
CAS Number:  
Certified Reference Material for Standardisation of Volumetric Solutions
Catalog # ACSDVS66332.80G
Supplier:  CPACHEM
CAS Number:  

Specifications

  • Pk:
    80 g
  • CAS No.:
    34962-29-3
  • GHS signal:
    Warning
  • Volume:
    80 g

Specifications

About this item

Iron(II)ethylenediammonium sulfate tetrahydrate - certified reference material for standardisation of volumetric solutions.

Shelf Life on Ship Date: 60 months - Store under normal laboratory conditions, at temperatures between 15 to 25 °C.

Certifications: ISO 17034, ISO 17025, ISO 9001.