Order Entry
Spain
ContactUsLinkComponent
Anti-Nectin-2/CD112 Rabbit Monoclonal Antibody [clone: S06-8F3]
Anti-Nectin-2/CD112 Rabbit Monoclonal Antibody [clone: S06-8F3]
Catalog # NOVUNBP3-20070-50
Supplier:  Novus Biologicals
Anti-Nectin-2/CD112 Rabbit Monoclonal Antibody [clone: S06-8F3]
Catalog # NOVUNBP3-20070-50
Supplier:  Novus Biologicals

Specifications

  • Antigen name:
    Poliovirus receptor related 2
  • Antigen symbol:
    Nectin-2/CD112
  • Clonality:
    Monoclonal
  • Clone:
    S06-8F3
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Antigen synonyms:
    HVEB|MPH|PRR2|PVRL2|Nectin2
  • Storage temperature:
    Store at 4 °C short term. Aliquot and store at −20 °C long term. Avoid freeze-thaw cycles.
  • Concentration:
    0,3 mg/ml
  • Shipping temperature:
    The product is shipped with polar packs. Upon receipt, store it immediately at the temperature recommended below.
  • Immunogen:
    A synthetic peptide of human Nectin-2/CD112 (Uniprot # Q92692)
  • Purification:
    Affinity purified
  • Pk:
    50 µl

Specifications

About this item

Nectin 2 encodes a single-pass type I membrane glycoprotein with two Ig-like C2-type domains and an Ig-like V-type domain. This protein is one of the plasma membrane components of adherens junctions.

  • Formulation: 50 mM tris-glycine (pH 7,4), 0,15 M NaCl, 40% glycerol, 0,05% BSA
  • Preservative: 0,01% sodium azide

It also serves as an entry for certain mutant strains of herpes simplex virus and pseudorabies virus, and it is involved in cell to cell spreading of these viruses. Variations in this gene have been associated with differences in the severity of multiple sclerosis. Alternate transcriptional splice variants, encoding different isoforms, have been characterised.

Type: Recombinant
Antigen: Nectin-2/CD112
Clonality: Monoclonal
Clone: S06-8F3
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat