Order Entry
Spain
ContactUsLinkComponent
Quanti-Cult™ Staphylococcus aureus subsp. aureus ATCC™ 25923™
Quanti-Cult™ Staphylococcus aureus subsp. aureus ATCC™ 25923™
Catalog # OXOIR4737010
CAS Number:  
Quanti-Cult™ Staphylococcus aureus subsp. aureus ATCC™ 25923™
Catalog # OXOIR4737010
CAS Number:  
Specifications
  • Pk:
    10
  • Description:
    Staphylococcus aureus subsp. aureus ATCC™ 25923™
Specifications
About this item
Quanti-Cult™ Staphylococcus aureus subsp. aureus ATCC™ 25923™ is supplied as a film of preserved microorganisms on the inside of the cap on a plastic vial.
  • No serial dilution required
  • Delivers <100 colony forming units (CFUs) in a single 0,1 ml inoculum
  • 10 tests/reconstituted vial; 100 tests/kit
  • Ready for rehydration
  • For use in quality control procedures
Packaging: Includes 10 preserved organism vials and 10 rehydration vials.