(±)-Isonipecotic acid ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:498-94-2
:
Formula:C₆H₁₁NO₂
MDL Number:MFCD00006004
:129.16 g/mol
Merck Index:14,5189
Boiling Pt:264 °C (1013 hPa)
Melting Pt:>350 °C
Storage Temperature:Ambient
EINECS:207-872-3
Related Information
Frequently Bought Together