Order Entry
Spain
ContactUsLinkComponent
Shoe Covers, PP Spunlaid Non Woven + PE Sole Plastic-Coated Sole
Shoe Covers, PP Spunlaid Non Woven + PE Sole Plastic-Coated Sole
Catalog # HOPE10.012LONG
Supplier:  Medicom
undefined
Shoe Covers, PP Spunlaid Non Woven + PE Sole Plastic-Coated Sole
Catalog # HOPE10.012LONG
Supplier:  Medicom

Specifications

  • Clothing Size:
    XX-Large
  • Garment application:
    Laboratory
  • Colour:
    White/blue
  • Description:
    Shoe covers
  • Pk:
    400
  • W×L:
    165×420 mm
  • Weight:
    30 g/m²

Specifications

About this item

Overshoes PP spunlaid non woven with PE sole Plastic-coated sole.

  • Overshoe with non-slip sole
  • They adapt to different morphologies and all types of shoes
  • Latex-free elastic
Certifications: ISO 9001 Manufacturing site.