

tert-Butyl ethyl ether for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:637-92-3
:
Formula:C₆H₁₄O
MDL Number:MFCD00009225
:102.18 g/mol
Merck Index:13,03809
Boiling Pt:71 °C (1013 hPa)
Melting Pt:–97 °C
Density:0.752 g/cm³ (20 °C)
UN:1179
ADR:3,II
:M-Clarity™ quality level = MQ200
EINECS:211-309-7
:GHS02
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:12 g/l (25 °C)
- Melting Point:-97 °C
- Molar Mass:102.18 g/mol
- Boiling Point:71 °C (1013 hPa)
- Vapor Pressure:173 hPa (25 °C)
- Flash Point:-19 °C
- Explosion Limit:1.23 - 7.7 %(V)
- Density:0.74 g/cm3 (20 °C)
- Hazard Class:3
- Hazard Class Description:Flammable Liquid
Specification Test Results
Frequently Bought Together