Methyl methacrylate stabilised for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:80-62-6
:
Formula:CH₂=C(CH₃)COOCH₃
MDL Number:MFCD00008587
:100.12 g/mol
Merck Index:12,06005
Boiling Pt:100 °C (1013 hPa)
Melting Pt:–48 °C
Density:0.93 g/cm³ (20 °C)
EINECS:201-297-1
UN:1247
ADR:3,II
:M-Clarity™ quality level = MQ200
Flash Pt:8 °C
REACH:01-2119452498-28
:GHS02
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:15 g/l (20 °C)
- Melting Point:-48 °C
- Molar Mass:100.12 g/mol
- Boiling Point:101 °C
- Vapor Pressure:53 hPa (20 °C)
- Flash Point:10 °C
- Refractive Index:1.414 (20 °C, 589 nm)
- Explosion Limit:2.1 - 12.5 %(V)
- Density:0.94 g/cm3 (20 °C)
- pH:(H2O) not applicable
- Ignition Point:430 °C
- Hazard Class:3
- Hazard Class Description:Flammable Liquid
Specification Test Results
Frequently Bought Together