Potassium dichromate 1/60 mol/l concentrated aqueous solution, Titrisol®, Supelco®
Catalog # 1.09928.0001
Supplier: Merck
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Plastic ampoule
- Pk:1 Ampoul
- Seq:0501
Specifications
Specification Test Results
- Solubility:(20 °C) soluble
- Density:1.06 g/cm3 (20 °C)
- pH:4.1 (H2O, 20 °C)
- Hazard Class:6.1
- Hazard Class Description:Toxic materials
Specification Test Results
About this item
:
:294.18 g/mol
ADR:6.1,
:M-Clarity™ quality level = MQ100
EINECS:231-906-6
UN:3287
:GHS06
:GHS05
:GHS08
:GHS09
Storage Temperature:Ambient
CAS Number:7778-50-9
Formula:K₂Cr₂O₇
MDL Number:MFCD00011367