Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Spain
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
L(+)-Potassium sodium tartrate
L(+)-Potassium sodium tartrate
L(+)-Potassium sodium tartrate
LOVI466101
:
Lovibond
:
L(+)-Potassium sodium tartrate
LOVI466101
:
Lovibond
:
Pk:
100 mL
Seq:
2440
:
:
210.16 g/mol
:
12,07840
:
90…100 °C
:
1.79 g/cm³ (20 °C)
:
206-156-8
:
304-59-6
:
C₄H₄KNaO₆
:
MFCD00065391