


Benzidine
Catalog # EHERC10536000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
:184.24 g/mol
Merck Index:12,01106
Boiling Pt:359 °C (1013 hPa)
Melting Pt:126…127 °C
Density:1.251 g/cm³ (20 °C)
EINECS:202-199-1
UN:1885
ADR:6.1,II
:GHS08
:GHS07
:GHS09
CAS Number:92-87-5
Formula:C₁₂H₁₂N₂
MDL Number:MFCD00039765