
Congo red ≥95%, Ultra Pure Grade
Catalog # AATB23056
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:696.67 g/mol
ADR:5,II
:GHS07
CAS Number:573-58-0
Formula:C₃₂H₂₂N₆Na₂O₆S₂
MDL Number:MFCD00004028