Order Entry
Spain
ContactUsLinkComponent
4-(4-(Dihexadecylamino)styryl)-N-methylpyridinium iodide (DiA) ≥95%
4-(4-(Dihexadecylamino)styryl)-N-methylpyridinium iodide (DiA) ≥95%
Catalog # AATB22030
Supplier:  AAT Bioquest
CAS Number:  
undefined
4-(4-(Dihexadecylamino)styryl)-N-methylpyridinium iodide (DiA) ≥95%
Catalog # AATB22030
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :787.04 g/mol
CAS Number:114041-00-8
Formula:C₄₆H₇₉N₂.I