Order Entry
Spain
Orders LinkContactUsLinkComponent
BAPTA tetrapotassium salt

BAPTA tetrapotassium salt

Plastic bottle, 100 mg
Catalog # AATB21003
Supplier:   AAT Bioquest
BAPTA tetrapotassium salt
Catalog # AATB21003
Supplier:  AAT Bioquest
CAS Number:  73630-08-7
BAPTA tetrapotassium salt
BAPTA tetrapotassium salt

Plastic bottle, 100 mg

CAS Number:  73630-08-7
Price
122,00 €
Log in for availability
1 * 100 mg
Product Details & Documents

BAPTA (1,2-bis(o-aminophenoxy)ethane-N,N,N',N'-tetraacetic acid) is a water-soluble and cell-impermeable calcium chelator.

Synonyms:  BAPTA 4K, 1,2-Bis(2-aminophenoxy)ethane-N,N,N′,N′-tetraacetic acid tetrapotassium salt, Potassium 2,2',2'',2'''-(((ethane-1,2-diylbis(oxy))bis(2,1-phenylene))bis(azanetriyl))tetraacetate
  • CAS Number:73630-08-7
  • Formula:C₂₂H₂₀N₂O₁₀K₄
  • MDL Number:MFCD00070080
  • Molecular Weight:628.79 g/mol
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mg

Specifications