Order Entry
Spain
Orders LinkContactUsLinkComponent
(Ac-LEHD)2-R110

(Ac-LEHD)2-R110

Plastic bottle, 1 mg
Catalog # AATB13427
Irritant
Supplier:   AAT Bioquest
(Ac-LEHD)2-R110
Catalog # AATB13427
Supplier:  AAT Bioquest
CAS Number:  
(Ac-LEHD)2-R110
(Ac-LEHD)2-R110

Plastic bottle, 1 mg

CAS Number:  
Price
156,00 €
Log in for availability
1 * 1 mg
Product Details & Documents

Caspase 9 is a member of the CED-3 subfamily of the caspase family of cysteine proteases that play an essential role in the execution phase of apoptosis.

  • Caspase detection
  • Bright signal
  • ADR:5,II
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.