Order Entry
Spain
ContactUsLinkComponent
Native Amanita phalloides Phalloidin (from Eukaryotic cells), Alexa Fluor® 532
Native Amanita phalloides Phalloidin (from Eukaryotic cells), Alexa Fluor® 532
Catalog # AATB23117
Supplier:  AAT Bioquest
CAS Number:  
undefined
Native Amanita phalloides Phalloidin (from Eukaryotic cells), Alexa Fluor® 532
Catalog # AATB23117
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    300 Tests
  • Conjugation:
    Alexa Fluor® 532
  • Protein/peptide type:
    Native
  • Source:
    Eukaryotic cells
  • Species:
    Amanita phalloides
  • Protein synonyms:
    Amanita phalloides
  • Protein/peptide name:
    Phalloidin

Specifications

About this item

This orange fluorescent phalloidin conjugate (equivalent to Alexa Fluor® 532-labeled phalloidin) selectively binds to F-actins.

  • Suitable for all fluorescent imaging platforms
  • Specific for actin in both living or fixed cells
  • Multiple applications
Caution: For research use only.