Helixyte™ Green *20X Aqueous PCR Solution*
Catalog # AATB17591
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pk:5 mL
- Description:Helixyte™ green
- Environmentally Preferable:
- Purity:95%
Specifications
About this item
Helixyte™ Gold is manufactured by AAT Bioquest, and it has the same chemical structure of SYBR® Gold (SYBR® is the trademark of ThermoFisher).
- Same spectral properties to SYBR® gold
- High sensitivity
- Useful for nucleic acid labeling in gels
Caution: For research use only.