Order Entry
Spain
ContactUsLinkComponent
Cell Meter™ Fluorimetric Intracellular Total ROS Activity Assay Kit*Orange Fluorescence*
Cell Meter™ Fluorimetric Intracellular Total ROS Activity Assay Kit*Orange Fluorescence*
Catalog # AATB22902
Supplier:  AAT Bioquest
CAS Number:  
undefined
Cell Meter™ Fluorimetric Intracellular Total ROS Activity Assay Kit*Orange Fluorescence*
Catalog # AATB22902
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pk:
    200 Tests
  • Description:
    Cell Meter™ fluorimetric intracellular total ROS activity assay kit*orange fluorescence*

Specifications

About this item

Reactive oxygen species (ROS) are natural byproducts of the normal metabolism of oxygen and play important roles in cell signaling.

  • Ready to use
  • Contains all necessary components
  • Robust and adaptable
  • Suitable for all major fluorescence platforms
  • Results with only 1 hour incubation
Caution: For research use only.