PhosphoWorks™ Luminometric ATP Assay Kit, Bright Glow
Supplier: AAT Bioquest
Certificates
About this item
Adenosine triphosphate (ATP) plays a fundamental role in cellular energenics, metabolic regulation and cellular signaling.
- Extreme sensitivity
- Detects as few as 10 cells/well
- No mixing or separations required
Caution: For research use only.
Frequently Bought Together