Cell Meter™ Caspase 9 Activity Apoptosis Assay Kit *Red Fluorescence*
Catalog # AATB22817
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pk:100 Tests
- Description:Cell Meter™ caspase 9 activity apoptosis assay kit *red fluorescence*
Specifications
About this item
Our Cell Meter™ assay kits are a set of tools for monitoring cellular functions.
- Contains all necessary components
- Multiple research applications
- Bright and stable signal
- Suitable for all major fluorescence platforms
Caution: For research use only.