Cl-NERF
1 mg
Catalog # AATB21220
Supplier: AAT Bioquest
Jump to:
Product Details & Documents
Cl-NERF is a rhodol dye with pKa = 3,8.
- pH-dependent fluorescence
- Ratiometric detection
- Cell impermeable
Caution: For research use only.
Specifications
Specifications
- Pk:1 mg