Order Entry
Spain
ContactUsLinkComponent
Native Streptomyces avidinii Streptavidin (from Streptomyces avidinii), IRDye® 800CW
Native Streptomyces avidinii Streptavidin (from Streptomyces avidinii), IRDye® 800CW
Catalog # AATB16979
Supplier:  AAT Bioquest
CAS Number:  
undefined
Native Streptomyces avidinii Streptavidin (from Streptomyces avidinii), IRDye® 800CW
Catalog # AATB16979
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    200 µG
  • Conjugation:
    IRDye® 800CW
  • Protein/peptide type:
    Native
  • Source:
    Streptomyces avidinii
  • Species:
    Streptomyces avidinii
  • Protein synonyms:
    Avidin|Streptomyces avidinii
  • Protein/peptide name:
    Streptavidin

Specifications

About this item

AAT Bioquest's iFluor® dyes are optimised for labeling proteins, in particular, antibodies.

  • Extremely high biotin-binding affinity
  • High quality conjugate
Caution: For research use only.