Order Entry
Spain
ContactUsLinkComponent
Cell Meter™ Phosphatidylserine Apoptosis Assay Kit *Deep Red Fluorescence Optimized for Flow Cytometry*
Cell Meter™ Phosphatidylserine Apoptosis Assay Kit *Deep Red Fluorescence Optimized for Flow Cytometry*
Catalog # AATB22832
Supplier:  AAT Bioquest
CAS Number:  
undefined
Cell Meter™ Phosphatidylserine Apoptosis Assay Kit *Deep Red Fluorescence Optimized for Flow Cytometry*
Catalog # AATB22832
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pk:
    100 Tests
  • Description:
    Cell Meter™ phosphatidylserine apoptosis assay kit *deep red fluorescence optimized for flow cytometry*
  • Tests per kit:
    100 Tests

Specifications

About this item

Our Cell Meter™ assay kits are a set of tools for monitoring cell viability.

  • Contains all necessary components
  • Multiple research applications
  • Bright and stable signal
  • Optimised for flow cytometry but suitable for all major fluorescence platforms
Caution: For research use only.