Cell Meter™ Live Cell Caspase 8 Binding Assay Kit *Red Fluorescence*
Catalog # AATB20116
Proveedor: AAT BIOQUEST
Número CAS:
Especificaciones
- Pk:25 Tests
- Description:Cell Meter™ Live cell Caspase 8 binding assay kit *Red fluorescence*
- Tests per kit:25 Tests
Especificaciones
Acerca de este artículo
Our Cell Meter™ live cell caspases activity assay kits are based on fluorescent FMK inhibitors of caspases.
- Based on FMK inhibitor
- Direct caspase detection
- Suitable for all major fluorescence platforms
Precaución: For research use only.