Order Entry
Spain
ContactUsLinkComponent
MycoLight™ Red JJ94 *2,5 mM in DMSO*
MycoLight™ Red JJ94 *2,5 mM in DMSO*
Catalog # AATB24006
Supplier:  AAT Bioquest
CAS Number:  
undefined
MycoLight™ Red JJ94 *2,5 mM in DMSO*
Catalog # AATB24006
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pk:
    100 µl
  • Description:
    MycoLight™ Red JJ94 stain
  • Purity:
    95%

Specifications

About this item

Green fluorescent protein (GFP) derivatives are widely used as fluorescent reporters to study biological processes at the cellular and molecular level, including labeling bacteria.

  • Cell-permeable
  • Minimal cytotoxicity
  • Useful for multicolour imaging
Caution: For research use only.