Goat Anti-Rabbit IgG (H&L) Agarose (10 mg)
Catalog # AATB55015
Supplier: AAT Bioquest
Specifications
- Antibody type:Secondary
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Clonality:Polyclonal
- Clone:IgG
- Conjugation:Agarose
- Host:Goat
- Isotype:IgG (H, L)
- Reactivity:Rabbit
- Size:10 mg
- Pack type:Vial
- Pk:10 mg
Specifications
About this item
Agarose conjugated antibodies are designed for the rapid and efficient collection of antibodies and proteins, including recombinant fusion proteins, from a complex protein mixture (see protocol).
- Excellent quality
- Affinity gel building block
- Multiple applications
Caution: For research use only.
Type: Secondary
Antigen: Immunoglobulin G
Clonality: Polyclonal
Clone: IgG
Conjugation: Agarose
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Rabbit