Order Entry
Spain
ContactUsLinkComponent
Biocytin hydrazide hydrochloride ≥95%
Irritant
Biocytin hydrazide hydrochloride ≥95%
Catalog # AATB3086
Supplier:  AAT Bioquest
CAS Number:  
undefined
Irritant
Biocytin hydrazide hydrochloride ≥95%
Catalog # AATB3086
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :422.97 g/mol
ADR:5,II
 :GHS07
CAS Number:102743-85-1
Formula:C₁₆H₃₁ClN₆O₃S