Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Spain
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Biotin amine
Biotin amine, ReadiView™ biodetection
Biotin amine, ReadiView™ biodetection
Irritant
Biotin amine, ReadiView™ biodetection
Catalog # AATB3053
Supplier:
AAT Bioquest
CAS Number:
Expand image
Irritant
Biotin amine, ReadiView™ biodetection
Catalog # AATB3053
Supplier:
AAT Bioquest
CAS Number:
Specifications
Pack type:
Plastic bottle
Pk:
5 mg
Seq:
2210
Specifications
About this item
Product Details
:
ADR:
5,II
:
GHS07
Product Details