Order Entry
Spain
ContactUsLinkComponent
 
Cyanine 7 bissuccinimidyl ester ≥90% [equivalent to Cy7® bisNHS ester]
  AATB170
 :  
undefined
Cyanine 7 bissuccinimidyl ester ≥90% [equivalent to Cy7® bisNHS ester]
  AATB170
 :  

 

  • Pk:
    1 mg
  • Seq:
    2070

 

 

 :
 :1064.27 g/mol