Order Entry
Spain
ContactUsLinkComponent
Cyanine 5 hydrazide ≥95% [equivalent to Cy5® hydrazide]
Cyanine 5 hydrazide ≥95% [equivalent to Cy5® hydrazide]
Catalog # AATB156
Supplier:  AAT Bioquest
CAS Number:  
undefined
Cyanine 5 hydrazide ≥95% [equivalent to Cy5® hydrazide]
Catalog # AATB156
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :898.89 g/mol