Order Entry
Spain
ContactUsLinkComponent
SPINeasy® Plasmid Purification, MiniPrep Kit, MP Biomedicals
SPINeasy® Plasmid Purification, MiniPrep Kit, MP Biomedicals
Catalog # 116534050.
Supplier:  MP Biomedicals
CAS Number:  
undefined
SPINeasy® Plasmid Purification, MiniPrep Kit, MP Biomedicals
Catalog # 116534050.
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Sample type:
    Bacterial culture
  • Target molecule:
    Plasmid
  • Description:
    SPINeasy plasmid MiniPrep kit, 50 tests
  • Storage temperature:
    Store at room Temperature (15...30 ℃), dry environment

Specifications

About this item

SPINeasy Plasmid Miniprep kit is a silica-membrane spin-column kit that enables up to 20 µg of plasmid DNA to be purified from 1 to 5 ml of transformed bacterial culture.

High purity plasmid DNA can be obtained within 25 minutes through a simple purification process and is immediately ready for routine molecular biology laboratory applications.

Key applications are In-vitro transcription, cloning, NGS, PCR, qPCR, restriction digestion, transformation, genomics, molecular biology.

Certifications: Hazard Class 8
Delivery: Kit includes 20 ml Resuspension Buffer SN1, 20 ml Alkaline Lysis Buffer SN2, 30 ml Neutralization Buffer SN3, 21 ml Wash Buffer NW1, 6 ml Wash Buffer NW2, 10 ml Elution Buffer SN, 200 µL RNase A, 50× Column SN with collection tubes.