Order Entry
Spain
ContactUsLinkComponent
Candida albicans IgM ELISA kit
Candida albicans IgM ELISA kit
Catalog # ABNOKA6723.1KIT
Supplier:  Abnova
CAS Number:  
Candida albicans IgM ELISA kit
Catalog # ABNOKA6723.1KIT
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Primary antibody reactivity:
    Candida albicans
  • Target protein:
    IgM
  • Assay Principle:
    Qualitative

Specifications

About this item

Candida albicans IgM ELISA Kit is a qualitative determination of IgM class antibodies against Candida albicans in human serum or plasma.