Recombinant Human PTPRC (from HEK293 cells), His-Avi tag
Catalog # ABNOP9751.100UG
Supplier: Abnova
CAS Number:
Specifications
- Pk:100 µG
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:His-Avi tag
- Gene ID:P08575-4
- Protein/peptide name:PTPRC