Skip to Header
Skip to Main Content
Skip to Footer
Avanzado
Avanzado
SDS
Certificados
Datos del fabricante
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Spain
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Recombinant Human TNFSF10 (from E. coli)
Recombinant Human TNFSF10 (from E. coli)
Recombinant Human TNFSF10 (from E. coli)
Catalog # ABNOP9292.50UG
Supplier:
Abnova
CAS Number:
Expand image
Recombinant Human TNFSF10 (from E. coli)
Catalog # ABNOP9292.50UG
Supplier:
Abnova
CAS Number:
Specifications
Pk:
50 µG
Protein/peptide type:
Recombinant
Source:
E. coli
Species:
Human
Protein/peptide name:
TNFSF10
Specifications
About this item
Product Details
Product Details