Order Entry
Spain
ContactUsLinkComponent
Recombinant Mouse TNFSF10 (from E. coli)
Recombinant Mouse TNFSF10 (from E. coli)
Catalog # ABNOP9295.50UG
Supplier:  Abnova
CAS Number:  
Recombinant Mouse TNFSF10 (from E. coli)
Catalog # ABNOP9295.50UG
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Protein/peptide name:
    TNFSF10

Specifications

About this item